COM448686951 Combi-Blocks, Inc.

methyl 3-(3-cyanophenyl)-2-methoxybenzoate

  • MDL number MFCD27578584
  • Cas Number : 1820608-76-1
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2005

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2005
Availability details for
Quantity
  • Availability
  • Add to Cart