COM448686953 Combi-Blocks, Inc.

methyl 2-chloro-4-(4-fluoro-3-methoxyphenyl)benzoate

  • MDL number MFCD27578586
  • Cas Number : 1820641-60-8
  • Molecular Weight : 294.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2009

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2009
Availability details for
Quantity
  • Availability
  • Add to Cart