COM448686956 Combi-Blocks, Inc.

methyl 2-chloro-4-[3-(trifluoromethoxy)phenyl]benzoate

  • MDL number MFCD27578588
  • Cas Number : 1820704-79-7
  • Molecular Weight : 330.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2012

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2012
Availability details for
Quantity
  • Availability
  • Add to Cart