COM448686962 Combi-Blocks, Inc.

methyl 3-(2-nitrophenyl)benzoate

  • MDL number MFCD27578591
  • Cas Number : 168619-44-1
  • Molecular Weight : 257.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2018

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2018
Availability details for
Quantity
  • Availability
  • Add to Cart