COM448686963 Combi-Blocks, Inc.

Methyl 2-(2,3-dihydro-1h-inden-5-yloxy)acetate

  • MDL number MFCD10692915
  • Cas Number : 1152514-86-7
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2019
Availability details for
Quantity
  • Availability
  • Add to Cart