COM448686965 Combi-Blocks, Inc.

2-Methoxy-3-(methoxycarbonyl)phenylboronic acid

  • MDL number MFCD23379502
  • Cas Number : 1332637-14-5
  • Molecular Weight : 209.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2021

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2021
Availability details for
Quantity
  • Availability
  • Add to Cart