COM448686969 Combi-Blocks, Inc.

methyl 4-(2-acetylphenyl)-2-chlorobenzoate

  • MDL number MFCD27578596
  • Cas Number : 1820711-85-0
  • Molecular Weight : 288.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2027
Availability details for
Quantity
  • Availability
  • Add to Cart