COM448686970 Combi-Blocks, Inc.

Methyl (4-chlorophenyl)(difluoro)acetate

  • MDL number MFCD22547876
  • Cas Number : 1393558-32-1
  • Molecular Weight : 220.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2029

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2029
Availability details for
Quantity
  • Availability
  • Add to Cart