COM448686971 Combi-Blocks, Inc.

1,3-dimethyl 5-[4-fluoro-3-(methoxycarbonyl)phenyl]benzene-1,3-dicarboxylate

  • MDL number MFCD27578597
  • Cas Number : 1820716-77-5
  • Molecular Weight : 346.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2032

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2032
Availability details for
Quantity
  • Availability
  • Add to Cart