COM448686974 Combi-Blocks, Inc.

methyl (2E)-3-[2-fluoro-3-(trifluoromethoxy)phenyl]prop-2-enoate

  • MDL number MFCD27578600
  • Cas Number : 1820749-05-0
  • Molecular Weight : 264.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2036

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2036
Availability details for
Quantity
  • Availability
  • Add to Cart