COM448686978 Combi-Blocks, Inc.

Methyl 5-[4-(benzyloxy)phenyl]nicotinate

  • MDL number MFCD06802748
  • Cas Number : 893740-27-7
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2040

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2040
Availability details for
Quantity
  • Availability
  • Add to Cart