COM448686987 Combi-Blocks, Inc.

methyl 2,2-difluoro-2-[2-(trifluoromethoxy)phenyl]acetate

  • MDL number MFCD27578610
  • Cas Number : 1820613-66-8
  • Molecular Weight : 270.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2055

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2055
Availability details for
Quantity
  • Availability
  • Add to Cart