COM448686991 Combi-Blocks, Inc.

methyl 2-fluoro-5-[3-(2-methoxy-2-oxoethyl)phenyl]benzoate

  • MDL number MFCD27578613
  • Cas Number : 1820674-15-4
  • Molecular Weight : 302.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2060

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2060
Availability details for
Quantity
  • Availability
  • Add to Cart