COM448686992 Combi-Blocks, Inc.

Methyl 2-[3-(trifluoromethyl)phenyl]benzoate

  • MDL number MFCD05979522
  • Cas Number : 773134-23-9
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2061
Availability details for
Quantity
  • Availability
  • Add to Cart