COM448686993 Combi-Blocks, Inc.

methyl 3-(4-fluorophenyl)pyridine-4-carboxylate

  • MDL number MFCD27578614
  • Cas Number : 873458-37-8
  • Molecular Weight : 231.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2062

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2062
Availability details for
Quantity
  • Availability
  • Add to Cart