COM448686997 Combi-Blocks, Inc.

2'-Hydroxymethyl-biphenyl-3-carboxylic acid methyl ester

  • MDL number MFCD05979052
  • Cas Number : 773872-89-2
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2071

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2071
Availability details for
Quantity
  • Availability
  • Add to Cart