COM448686999 Combi-Blocks, Inc.

Methyl 5-(4-methylphenyl)nicotinate

  • MDL number MFCD06802206
  • Cas Number : 893735-14-3
  • Molecular Weight : 227.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2073
Availability details for
Quantity
  • Availability
  • Add to Cart