COM448687003 Combi-Blocks, Inc.

Methyl 2-[2-(hydroxymethyl)phenyl]benzoate

  • MDL number MFCD05980276
  • Cas Number : 38399-64-3
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2077

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2077
Availability details for
Quantity
  • Availability
  • Add to Cart