COM448687004 Combi-Blocks, Inc.

methyl 3-(4-methoxyphenyl)pyridine-4-carboxylate

  • MDL number MFCD27578621
  • Cas Number : 1820613-47-5
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2078
Availability details for
Quantity
  • Availability
  • Add to Cart