COM448687006 Combi-Blocks, Inc.

Methyl 4-[(dimethylamino)sulfonyl]benzoate

  • MDL number MFCD01113321
  • Cas Number : 58804-17-4
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2080
Availability details for
Quantity
  • Availability
  • Add to Cart