COM448687007 Combi-Blocks, Inc.

methyl 1-(2-fluorophenyl)cyclopentane-1-carboxylate

  • MDL number MFCD27578622
  • Cas Number : 943119-08-2
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2081

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2081
Availability details for
Quantity
  • Availability
  • Add to Cart