COM448687010 Combi-Blocks, Inc.

Methyl 3-nitro-5-phenylbenzoate

  • MDL number MFCD25710851
  • Cas Number : 458537-98-9
  • Molecular Weight : 257.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2085

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2085
Availability details for
Quantity
  • Availability
  • Add to Cart