COM448687011 Combi-Blocks, Inc.

methyl 3-[2-(trifluoromethoxy)phenyl]benzoate

  • MDL number MFCD27578624
  • Cas Number : 1820613-75-9
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2086

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2086
Availability details for
Quantity
  • Availability
  • Add to Cart