COM448687013 Combi-Blocks, Inc.

methyl 2-[4-(3-chloro-5-fluorophenyl)phenyl]acetate

  • MDL number MFCD27578626
  • Cas Number : 1820613-54-4
  • Molecular Weight : 278.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2090
Availability details for
Quantity
  • Availability
  • Add to Cart