COM448687014 Combi-Blocks, Inc.

methyl 2-(4-{4-[(piperidin-1-yl)carbonyl]phenyl}phenyl)acetate

  • MDL number MFCD27578627
  • Cas Number : 1820684-65-8
  • Molecular Weight : 337.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2091

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2091
Availability details for
Quantity
  • Availability
  • Add to Cart