COM448687015 Combi-Blocks, Inc.

methyl 5-(4-tert-butylphenyl)-2-chlorobenzoate

  • MDL number MFCD27578628
  • Cas Number : 1820705-07-4
  • Molecular Weight : 302.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2093

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2093
Availability details for
Quantity
  • Availability
  • Add to Cart