COM448687018 Combi-Blocks, Inc.

Methyl 2'-acetyl[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD06801760
  • Cas Number : 893734-80-0
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2098
Availability details for
Quantity
  • Availability
  • Add to Cart