COM448687022 Combi-Blocks, Inc.

[3-(1-methoxy-2-methyl-1-oxopropan-2-yl)phenyl]boronic acid

  • MDL number MFCD27578633
  • Cas Number : 2246658-07-9
  • Molecular Weight : 222.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2103

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2103
Availability details for
Quantity
  • Availability
  • Add to Cart