COM448687026 Combi-Blocks, Inc.

methyl 5-(3-chlorophenyl)-2-fluorobenzoate

  • MDL number MFCD27578637
  • Cas Number : 1820740-96-2
  • Molecular Weight : 264.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2109

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2109
Availability details for
Quantity
  • Availability
  • Add to Cart