COM448687030 Combi-Blocks, Inc.

methyl 6-(3-fluorophenyl)pyridine-3-carboxylate

  • MDL number MFCD27578640
  • Cas Number : 1820609-01-5
  • Molecular Weight : 231.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2115
Availability details for
Quantity
  • Availability
  • Add to Cart