COM448687032 Combi-Blocks, Inc.

methyl 2-{3-[4-(trifluoromethoxy)phenyl]phenyl}acetate

  • MDL number MFCD27578642
  • Cas Number : 1820735-92-9
  • Molecular Weight : 310.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2117

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2117
Availability details for
Quantity
  • Availability
  • Add to Cart