COM448687035 Combi-Blocks, Inc.

methyl 4-(2-aminophenyl)benzoate hydrochloride

  • MDL number MFCD27578644
  • Cas Number : 1820741-08-9
  • Molecular Weight : 263.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2122

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2122
Availability details for
Quantity
  • Availability
  • Add to Cart