COM448687036 Combi-Blocks, Inc.

Methyl 5-([(tert-butoxy)carbonyl]amino)-2-methoxybenzoate

  • MDL number MFCD24468004
  • Cas Number : 180690-86-2
  • Molecular Weight : 281.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2125

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2125
Availability details for
Quantity
  • Availability
  • Add to Cart