COM448687037 Combi-Blocks, Inc.

methyl 5-(4-tert-butylphenyl)pyridine-2-carboxylate

  • MDL number MFCD27578645
  • Cas Number : 1820619-54-2
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2126

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2126
Availability details for
Quantity
  • Availability
  • Add to Cart