COM448687039 Combi-Blocks, Inc.

methyl 3-[4-(methylsulfanyl)phenyl]-5-(trifluoromethyl)benzoate

  • MDL number MFCD27578647
  • Cas Number : 1820716-50-4
  • Molecular Weight : 326.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2128

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2128
Availability details for
Quantity
  • Availability
  • Add to Cart