COM448687040 Combi-Blocks, Inc.

methyl (2S,3R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(triphenylmethoxy)butanoate

  • MDL number MFCD27578648
  • Cas Number : 1220123-46-5
  • Molecular Weight : 597.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2129

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2129
Availability details for
Quantity
  • Availability
  • Add to Cart