COM448687044 Combi-Blocks, Inc.

[2-fluoro-6-(methoxycarbonyl)pyridin-3-yl]boronic acid

  • MDL number MFCD27578649
  • Cas Number : 2225177-42-2
  • Molecular Weight : 198.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2133

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2133
Availability details for
Quantity
  • Availability
  • Add to Cart