COM448687045 Combi-Blocks, Inc.

Methyl 4'-acetyl[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD06204214
  • Cas Number : 525362-25-8
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2134
Availability details for
Quantity
  • Availability
  • Add to Cart