COM448687047 Combi-Blocks, Inc.

methyl 2-hydroxy-4-[4-(methoxycarbonyl)phenyl]benzoate

  • MDL number MFCD27578650
  • Cas Number : 848155-29-3
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2137

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2137
Availability details for
Quantity
  • Availability
  • Add to Cart