COM448687050 Combi-Blocks, Inc.

Methyl 2'-fluoro-1,1'-biphenyl-4-carboxylate

  • MDL number MFCD00662461
  • Cas Number : 80254-85-9
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2140

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2140
Availability details for
Quantity
  • Availability
  • Add to Cart