COM448687054 Combi-Blocks, Inc.

methyl 2-fluoro-5-(4-fluoro-2-methylphenyl)benzoate

  • MDL number MFCD27578654
  • Cas Number : 1820620-10-7
  • Molecular Weight : 262.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2145
Availability details for
Quantity
  • Availability
  • Add to Cart