COM448687056 Combi-Blocks, Inc.

methyl 4-hydroxy-6-methylquinoline-3-carboxylate

  • MDL number MFCD27578656
  • Cas Number : 942870-88-4
  • Molecular Weight : 217.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2147

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2147
Availability details for
Quantity
  • Availability
  • Add to Cart