COM448687058 Combi-Blocks, Inc.

methyl 3-chloro-5-[4-(hydroxymethyl)phenyl]benzoate

  • MDL number MFCD27578657
  • Cas Number : 1820683-81-5
  • Molecular Weight : 276.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2150
Availability details for
Quantity
  • Availability
  • Add to Cart