COM448687060 Combi-Blocks, Inc.

methyl 4-(4-tert-butylphenyl)-2-fluorobenzoate

  • MDL number MFCD27578659
  • Cas Number : 1703798-86-0
  • Molecular Weight : 286.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2153
Availability details for
Quantity
  • Availability
  • Add to Cart