COM448687063 Combi-Blocks, Inc.

methyl 3-[4-(trifluoromethoxy)phenyl]benzoate

  • MDL number MFCD27578662
  • Cas Number : 1820666-93-0
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2157

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2157
Availability details for
Quantity
  • Availability
  • Add to Cart