COM448687066 Combi-Blocks, Inc.

methyl 3-phenyl-5-(trifluoromethyl)benzoate

  • MDL number MFCD27578665
  • Cas Number : 101640-74-8
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2160

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2160
Availability details for
Quantity
  • Availability
  • Add to Cart