COM448687071 Combi-Blocks, Inc.

4'-Fluoro-3-hydroxy-[1,1'-biphenyl]-4-carboxylic acid methyl ester

  • MDL number MFCD14525661
  • Cas Number : 873784-26-0
  • Molecular Weight : 246.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2166

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2166
Availability details for
Quantity
  • Availability
  • Add to Cart