COM448687072 Combi-Blocks, Inc.

methyl 3-(diethoxyphosphoryl)propanoate

  • MDL number MFCD00137949
  • Cas Number : 1112-94-3
  • Molecular Weight : 224.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2168

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2168
Availability details for
Quantity
  • Availability
  • Add to Cart