COM448687080 Combi-Blocks, Inc.

methyl 2-[2-(methylsulfanyl)phenyl]benzoate

  • MDL number MFCD27578675
  • Cas Number : 1372768-61-0
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2177

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2177
Availability details for
Quantity
  • Availability
  • Add to Cart