COM448687086 Combi-Blocks, Inc.

methyl 3-[3-fluoro-5-(methoxycarbonyl)phenyl]-5-nitrobenzoate

  • MDL number MFCD27578680
  • Cas Number : 1820711-32-7
  • Molecular Weight : 333.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2185

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-2185
Availability details for
Quantity
  • Availability
  • Add to Cart